EMAIL THIS PAGE TO A FRIEND
B4027 Sigma-Aldrich

Bucetin

analytical standard

Synonym: 3-Hydroxy-p-butyrophenetidide, N-(4-Ethoxyphenyl)-3-hydroxybutanamide

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Opiates / Synthetic Analgesic Drug Standards, Pharmaceuticals, Illicit Drugs & Alcohol
grade   analytical standard
InChI Key   LIAWQASKBFCRNR-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Packaging

10 g in poly bottle

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
EU0900000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?