• USA Home
  • B5650 - Bephenium hydroxynaphthoate

EMAIL THIS PAGE TO A FRIEND
B5650 Sigma-Aldrich

Bephenium hydroxynaphthoate

analytical standard

Synonym: Benzyldimethyl(2-phenoxyethyl)ammonium 3-hydroxynaphthoate

Purchase

Properties

Related Categories Additional Drugs, Additional Standards, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
InChI Key   PMPQCPQAHTXCDK-UHFFFAOYSA-M
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Packaging

10 g in poly bottle

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?