• USA Home
  • B6131 - Bromophenol Blue sodium salt

EMAIL THIS PAGE TO A FRIEND
B6131 Sigma-Aldrich

Bromophenol Blue sodium salt

Technical grade

Synonym: 3′,3′′,5′,5′′-Tetrabromophenolsulfophthalein sodium salt

  • CAS Number 34725-61-6

  • Empirical Formula (Hill Notation) C19H9Br4NaO5S

  • Molecular Weight 691.94

  •  Beilstein Registry Number 6995701

  •  EC Number 252-170-2

  •  MDL number MFCD00013793

  •  PubChem Substance ID 329773271

Purchase

Properties

Related Categories B, Cell Biology, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   TWKDJMRHSLWRFI-UHFFFAOYSA-M
suitability   suitable for histology

Description

Application

A tracking dye for nucleic acid or protein electrophoresis in agarose or polyacrylamide gels.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?