EMAIL THIS PAGE TO A FRIEND
B8136 Sigma-Aldrich

Bromosulfonazo III

Synonym: 3,6-Bis(4-bromo-2-sulfophenylazo)-4,5-dihydroxy-2,7-naphthalenedisulfonic acid, 3,6-bis(4-Bromo-2-sulfophenylazo)-4,5-dihydroxy-2,7-naphthalenedisulfonic acid

Purchase

Properties

Related Categories B, Cell Biology, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
composition   Dye content, ~80%

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?