EMAIL THIS PAGE TO A FRIEND
B8767 Sigma-Aldrich

Barium acetate

powder

  • CAS Number 543-80-6

  • Linear Formula (CH3COO)2Ba

  • Molecular Weight 255.42

  •  Beilstein Registry Number 3693411

  •  EC Number 208-849-0

  •  MDL number MFCD00012447

Purchase

Properties

Related Categories Barium, Barium Salts, Chemical Synthesis, Essential Chemicals, Inorganic Salts,
InChI Key   ITHZDDVSAWDQPZ-UHFFFAOYSA-L
form   powder
color   white

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
AF4550000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?