• USA Home
  • B8900 - Benzoylecgonine solution

EMAIL THIS PAGE TO A FRIEND
B8900 Sigma-Aldrich

Benzoylecgonine solution

analytical standard, 1.0 mg/mL in methanol

Purchase

Properties

Related Categories Additional Standards, Alkaloids, Analytical Standards, Analytical/Chromatography, Chemical Structure,
grade   analytical standard
InChI Key   GVGYEFKIHJTNQZ-RFQIPJPRSA-N
drug control   Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschland
concentration   1.0 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C
Gene Information   rat ... Slc6a3(24898)

Description

Packaging

1 mL in ampule

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Protocols & Articles

Articles

HPLC Analysis of Drugs of Abuse on Ascentis® Express F5

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Drugs of Abuse on Ascentis® Express F5
Keywords: Chromatography, High performance liquid chromatography

LC/MS Analysis of Illicit Drugs and Metabolites on Ascentis® Express RP-Amide

From our library of Articles, Sigma-Aldrich presents LC/MS Analysis of Illicit Drugs and Metabolites on Ascentis® Express RP-Amide
Keywords: Chromatography, Forensic, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Separation

LC/MS/MS Analysis of Drugs of Abuse: Generic Screening on Ascentis® Express F5

From our library of Articles, Sigma-Aldrich presents LC/MS/MS Analysis of Drugs of Abuse: Generic Screening on Ascentis® Express F5
Keywords: Chromatography, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Solvents

UHPLC/MS/MS Analysis of 25 Small Molecule Pharmaceutical Compounds on Ascentis® Express RP-Amide

From our library of Articles, Sigma-Aldrich presents UHPLC/MS/MS Analysis of 25 Small Molecule Pharmaceutical Compounds on Ascentis® Express RP-Amide
Keywords: Chromatography, High performance liquid chromatography, Mass spectrometry, Pharmaceutical, Separation

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?