• USA Home
  • B9876 - Sodium tetraborate decahydrate

EMAIL THIS PAGE TO A FRIEND
B9876 Sigma-Aldrich

Sodium tetraborate decahydrate

ReagentPlus®, ≥99.5%

Synonym: Borax decahydrate, Sodium borate decahydrate

Purchase

Properties

Related Categories Essential Chemicals, Research Essentials, Salts, Sodium Salts
grade   ReagentPlus®
assay   ≥99.5%
density   1.73 g/mL at 25 °C(lit.)

Description

Application

Widely used as a buffer. pH of a solution is approx. 9.3 over a wide range of concentrations.

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Packaging

1 kg in poly bottle

5 kg in poly drum

500 g in poly bottle

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
1
RTECS 
VZ2275000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?