EMAIL THIS PAGE TO A FRIEND
BCR017A Sigma-Aldrich

Copper (S, P)

BCR® certified Reference Material, disc

Synonym: Copper

Purchase

Description

Analysis Note

For more information please see:
BCR017A

General description

Certified for the mass fractions of phosphorus and sulfur.

Legal Information

BCR is a registered trademark of European Commission

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN3089 - class 4.1 - PG 2 - EHS - Metal powders, flammable, n.o.s., Hi:A,S
WGK Germany 
3
RTECS 
GL5325000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?