• USA Home
  • BCR022B - Electrolytic tough pitch copper (O)

EMAIL THIS PAGE TO A FRIEND
BCR022B Sigma-Aldrich

Electrolytic tough pitch copper (O)

BCR® certified Reference Material, rod

Synonym: Copper

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application CRMs, Certified Reference Materials (CRMs),
grade   BCR® certified Reference Material
InChI Key   RYGMFSIKBFXOCR-UHFFFAOYSA-N
form   rod
resistivity   1.673 μΩ-cm, 20°C
bp   2567 °C(lit.)
mp   1083.4 °C(lit.)
density   8.94 g/mL at 25 °C(lit.)
format   matrix material

Description

Analysis Note

For more information please see:
BCR022B

General description

Certified for the mass fractions of oxygen.

Legal Information

BCR is a registered trademark of European Commission

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?