• USA Home
  • BCR047 - Benzo[b]fluoranthene

EMAIL THIS PAGE TO A FRIEND
BCR047 Sigma-Aldrich

Benzo[b]fluoranthene

BCR® certified Reference Material

Synonym: 2,3-Benzfluoranthene, 3,4-Benzofluoranthene, 3,4-Benz[e]acephenanthrylene, 3,4-Benzfluoranthene, Benz[e]acephenanthrylene, Benzo[e]fluoranthene, NSC 89265

Purchase

Description

Analysis Note

For more information please see:
BCR047

Legal Information

BCR is a registered trademark of European Commission

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?