• USA Home
  • BCR090B - Titanium (impurities)
EMAIL THIS PAGE TO A FRIEND
BCR090B Sigma-Aldrich
Titanium (impurities)
BCR® certified Reference Material, cubes (0.2 g)
Purchase
Description
Analysis Note
For more information please see:
BCR090B
General description
Certified for the mass fractions of B, Co, Cr, Cu, Fe, Mn, Mo and Ni.
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Legal Information
BCR is a registered trademark of European Commission
Price and Availability
Suggested Laboratory Gloves
Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
RTECS 
XR1700000
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?