• USA Home
  • BCR094 - Dibenz[a,c]anthracene

EMAIL THIS PAGE TO A FRIEND
BCR094 Sigma-Aldrich

Dibenz[a,c]anthracene

BCR® certified Reference Material

Synonym: 1,2:3,4-Dibenzanthracene

  • CAS Number 215-58-7

  • Empirical Formula (Hill Notation) C22H14

  • Molecular Weight 278.35

  •  Beilstein Registry Number 1912415

  •  EC Number 205-920-8

  •  MDL number MFCD00003603

  •  PubChem Substance ID 329774218

Purchase

Description

Analysis Note

For more information please see:
BCR094

Legal Information

BCR is a registered trademark of European Commission

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 3 - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
DM1925000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?