• USA Home
  • BCR123A - Reference ethanols (H, M, L) (for SNIF-NMR)

EMAIL THIS PAGE TO A FRIEND
BCR123A Sigma-Aldrich

Reference ethanols (H, M, L) (for SNIF-NMR)

BCR® certified Reference Material, tubes

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application CRMs, Certified Reference Materials (CRMs),
grade   BCR® certified Reference Material
form   tubes
diam.   10 mm

Description

Analysis Note

For more information please see:
BCR123A

Legal Information

BCR is a registered trademark of European Commission

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?