EMAIL THIS PAGE TO A FRIEND
BCR154 Sigma-Aldrich

Dibenz[a,j]acridine

BCR® certified Reference Material

Synonym: 7-Azadibenz[a,j]anthracene

  • CAS Number 224-42-0

  • Empirical Formula (Hill Notation) C21H13N

  • Molecular Weight 279.33

  •  Beilstein Registry Number 18508

  •  MDL number MFCD00055947

  •  PubChem Substance ID 329774232

Purchase

Description

Analysis Note

For more information please see:
BCR154

Legal Information

BCR is a registered trademark of European Commission

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
HN1050000

Documents

Certificate of Analysis

Protocols & Articles

Articles

US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608

From our library of Articles, Sigma-Aldrich presents US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?