• USA Home
  • BCR286A - Electrolytically refined lead (trace elements)

EMAIL THIS PAGE TO A FRIEND
BCR286A Sigma-Aldrich

Electrolytically refined lead (trace elements)

BCR® certified Reference Material, square blocks

Purchase

Description

Analysis Note

For more information please see:
BCR286A

General description

Certified for the mass fractions of Bi, Sb, Tl and Zn.

Legal Information

BCR is a registered trademark of European Commission

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
nwg
RTECS 
OF7525000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?