• USA Home
  • BCR446 - Rapeseed (oil, moisture, volatiles)

EMAIL THIS PAGE TO A FRIEND
BCR446 Sigma-Aldrich

Rapeseed (oil, moisture, volatiles)

BCR® certified Reference Material

Purchase

Description

Analysis Note

For more information please see:
BCR446

Application

The rapeseed reference material is to be analyzed according to the standard solvent procedure EEC, FOSFA or ISO. It can be used to calibrate NMR methods for the analysis of seeds of the same variety.
Mass fraction (g/100 g):
′As is’ Oil : 39.49 ± 0.14; Moisture and volatiles 7.01 ± 0.07; Dry basis Oil 42.48 ± 0.15

Packaging

About 150 g of rapeseed in a specially laminated bag containing an oxygen absorber and sealed under dry argon.

Legal Information

BCR is a registered trademark of European Commission

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?