• USA Home
  • BCR538 - Plastic film (overall migration in olive oil, film B)
EMAIL THIS PAGE TO A FRIEND
BCR538 Sigma-Aldrich
Plastic film (overall migration in olive oil, film B)
BCR® certified Reference Material
Purchase
Description
Analysis Note
For more information please see:
BCR-538_cert
BCR-538_report
Application
This CRM consists of additive free linear low density polyethylene sheets. It is intended to determine the overall migration applying the draft ENV 1186 standard procedure established by the Technical Committee 194 of the European Committee for Standardisation (CEN).
Overall migration by single cell in olive oil 10 days at 40°C : 5.7 ± 0.7 (mg/dm2)
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Packaging
PET/foil/PE heat sealed pouche containing double thickness sheets of additive free linear low densitiy polyethylene of 125x22.5 cm
Legal Information
BCR is a registered trademark of European Commission
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
08168 Timestrip Plus 8 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?