• USA Home
  • BCR564 - SiO2/Si (10, 20, 30 nm)
EMAIL THIS PAGE TO A FRIEND
BCR564 Sigma-Aldrich
SiO2/Si (10, 20, 30 nm)
BCR® certified Reference Material
Purchase
Description
Analysis Note
For more information please see:
BCR-564_cert
BCR-564_report
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Packaging
Set of three standards (16x16 mm): 1 standard each of 10, 50 and 120 nm SiO2 on silicon wafer.
Legal Information
BCR is a registered trademark of European Commission
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?