• USA Home
  • BCR626 - Arsenobetaine Standard solution

EMAIL THIS PAGE TO A FRIEND
BCR626 Sigma-Aldrich

Arsenobetaine Standard solution

BCR® certified Reference Material

Synonym: Arsenobetaine solution

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application CRMs, Certified Reference Materials (CRMs),
grade   BCR® certified Reference Material
InChI Key   SPTHHTGLGVZZRH-UHFFFAOYSA-N
format   single component solution

Description

Legal Information

BCR is a registered trademark of European Commission

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?