EMAIL THIS PAGE TO A FRIEND
C0415 Sigma-Aldrich

Ciclopirox olamine

analytical standard

Synonym: 6-Cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridone ethanolammonium salt

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Antifungals Standards, Chemical Structure,
grade   analytical standard
InChI Key   MBRHNTMUYWQHMR-UHFFFAOYSA-N
assay   ≥97%
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Packaging

1, 5 g in poly bottle

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
UU7785500

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?