CAS Number 10043-52-4
Linear Formula CaCl2
Molecular Weight 110.98
EC Number 233-140-8
MDL number MFCD00010903
PubChem Substance ID 24892300
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Apoptosis and Cell Cycle, Buffers, Calcium, Cell Biology, Cell Signaling and Neuroscience, |
| grade | anhydrous |
| vapor pressure | 0.01 mmHg ( 20 °C) |
| assay | ≥93.0% |
| form | granular |
| particle size | ≤7.0 mm |
| mp | 772 °C(lit.) |
may contain powder and some dark grey granules
100, 500 g in poly bottle
2.5 kg in poly drum
Customers Also Viewed

anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?