• USA Home
  • C139 - Cocaine propyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
C139 Sigma-Aldrich

Cocaine propyl ester hydrochloride

analytical standard

Purchase

Properties

Related Categories Additional Standards, Alkaloids, Analytical Standards, Analytical/Chromatography, Chemical Structure,
grade   analytical standard
InChI Key   ROEIOKRMKFGKTA-RGWPEEHPSA-N
form   neat
drug control   USDEA Schedule II; Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 1544PSN2 6.1 / PGII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?