• USA Home
  • C150 - Desmethyl-4′-chlorodiazepam

EMAIL THIS PAGE TO A FRIEND
C150 Sigma-Aldrich

Desmethyl-4′-chlorodiazepam

analytical standard, >96%, solid

Synonym: 7-Chloro-5-(4-chlorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one, Desmethyl Ro 05-4864

  • Empirical Formula (Hill Notation) C15H10Cl2N2O

  • Molecular Weight 305.16

  •  MDL number MFCD00153781

  •  PubChem Substance ID 24892415

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Benzodiazepin Analoga Structure, Benzodiazepines Standards,
grade   analytical standard
InChI Key   XKAHDJVCUSNHTE-UHFFFAOYSA-N
assay   >96%
form   neat
  solid
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
color   light tan
solubility   H2O: insoluble
  ethanol: soluble
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Packaging

5 mg in glass bottle

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?