EMAIL THIS PAGE TO A FRIEND
C2290 Sigma-Aldrich

Clofoctol

analytical standard

Synonym: 2-(2,4-Dichlorobenzyl)-4-(1,1,3,3-tetramethylbutyl)phenol

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Antibiotics Standards, Chromatography,
grade   analytical standard
InChI Key   HQVZOORKDNCGCK-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Packaging

1 g in poly bottle

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. C2290.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?