C2432 Sigma-Aldrich

Chloroform

contains 100-200 ppm amylenes as stabilizer, ≥99.5%

DOWNLOAD MSDS (PDF)

Synonym: Methylidyne trichloride, Trichloromethane

Description

Frequently Asked Questions

Live Chat and Frequently Asked Questions are available for this Product.

Packaging

1, 2.5 L in glass bottle

25, 500 mL in glass bottle

Application

Chromatin isolation by RNA purification (ChIRP). Ci C Chu, Jeffrey J Quinn, Howard Y HY Chang, Journal of Visualized Experiments 2012-01-01
Chromatin isolation by RNA purification (ChIRP)


Genomic maps of long noncoding RNA occupancy reveal principles of RNA-chromatin interactions.
Ci C Chu, Kun K Qu, Franklin L FL Zhong, Steven E SE Artandi, Howard Y HY Chang, Molecular Cell 2011-11-18
Genomic maps of long noncoding RNA occupancy reveal principles of RNA-chromatin interactions

Price and Availability

Customers Also Viewed

2-Propanol

BioReagent, for molecular biology, ≥99%

Chloroform

≥99%, PCR Reagent, contains amylenes as stabilizer

Chloroform

anhydrous, contains amylenes as stabilizer, ≥99%

Chloroform

anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer

Chloroform

CHROMASOLV® Plus, for HPLC, ≥99.9%, contains amylenes as stabilizer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?