• USA Home
  • C3141 - Cadmium chloride hemi(pentahydrate)

EMAIL THIS PAGE TO A FRIEND
C3141 Sigma-Aldrich

Cadmium chloride hemi(pentahydrate)

≥98% (titration)

Purchase

Properties

Related Categories Cadmium, Cadmium Salts, Cancer Research, Carcinogens, Catalysis and Inorganic Chemistry,
vapor density   6.3 (vs air)
InChI Key   DZVRGWYMCGLNKJ-UHFFFAOYSA-J
assay   ≥98% (titration)

Description

Biochem/physiol Actions

Carcinogen that acts by inhibiting DNA mismatch repair. Environmental pollutant.

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 2570 6.1 / PGIII
WGK Germany 
3
RTECS 
EV0178000
Protocols & Articles

Related Content

Carcinogen Selector - Reproductive System - Prostate

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Reproductive System - Prostate
Keywords: Cancer, Carcinogens, Respiratory

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?