CAS Number 10101-41-4
Linear Formula CaSO4 · 2H2O
Molecular Weight 172.17
EC Number 231-900-3
MDL number MFCD00149625
PubChem Substance ID 24892640
Popular Documents: Product Information Sheet (PDF)
| Related Categories | Calcium, Chemical Synthesis, Inorganic Salts, Synthetic Reagents |
| grade | ReagentPlus® |
| assay | ≥99% |
1 kg in poly bottle
500 g in poly bottle
ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?