CAS Number 10035-04-8
Empirical Formula (Hill Notation) CaCl2 · 2H2O
Molecular Weight 147.01
EC Number 233-140-8
MDL number MFCD00149613
PubChem Substance ID 24892650
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| grade | ReagentPlus® |
| assay | ≥99.0% |
| mp | 176 °C (dec.)(lit.) |
1 kg in poly bottle
2.5, 5, 12 kg in poly drum
500 g in poly bottle
ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?