• USA Home
  • C3929 - Collagen from kangaroo tail

EMAIL THIS PAGE TO A FRIEND
C3929 Sigma-Aldrich

Collagen from kangaroo tail

Bornstein and Traub Type I, powder

Purchase

Properties

Related Categories Biochemicals and Reagents, Collagen, Proteins and Derivatives, Structural Proteins
form   powder
solubility   : soluble (Clear to slightly hazy colorless solution at 1 mg/mL in water plus 2μL of acetic acid)
storage temp.   2-8°C

Description

General description

Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub. Do not confuse Sigma type designations with recognized collagen classification types.

Preparation Note

Prepared by a modification of the pepsin extraction and salt precipitation method of Niyibizi, et al., J. Biol. Chem., 259, 14170 (1984).

Application

Collagen Type I has been used as a scaffold for the growth in vitro of stem cells in a wide variety of biomaterial engineering studies.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?