• USA Home
  • C4665 - Camylofine dihydrochloride

EMAIL THIS PAGE TO A FRIEND
C4665 Sigma-Aldrich

Camylofine dihydrochloride

analytical standard, for drug analysis

Synonym: α-([Diethylaminoethyl]amino)phenylacetic acid isopentyl ester

Purchase

Properties

Related Categories Additional Drugs, Additional Standards, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard, for drug analysis
InChI Key   ZHRVNCCPAKIGRH-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Packaging

1 g in poly bottle

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
MB9625000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?