• USA Home
  • C7153 - 5(6)-Carboxyfluorescein Mixed isomers

EMAIL THIS PAGE TO A FRIEND
C7153 Sigma-Aldrich

5(6)-Carboxyfluorescein Mixed isomers

~99% (HPLC)

Purchase

Description

Other Notes

A probe for liposome-cell interactions.

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?