Synonym: Blue Vitriol, Cupric sulfate pentahydrate, Sulfuric acid, copper(II) salt (1:1) pentahydrate
CAS Number 7758-99-8
Linear Formula CuSO4 · 5H2O
Molecular Weight 249.69
EC Number 231-847-6
MDL number MFCD00149681
PubChem Substance ID 24892976
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Essential Chemicals, Inorganic Salts, Solutions and Reagents, Reagent Plus, Research Essentials |
| grade | ReagentPlus® |
| vapor pressure | 7.3 mmHg ( 25 °C) |
| assay | ≥98.0% |
| mp | 110 °C (dec.)(lit.) |
1 kg in poly bottle
250, 500 g in poly bottle
ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?