CAS Number 1305-62-0
Linear Formula Ca(OH)2
Molecular Weight 74.09
EC Number 215-137-3
MDL number MFCD00010901
Popular Documents: Specification Sheet (PDF)
| Related Categories | Chemical Synthesis, Inorganic Bases, Synthetic Reagents More... |
| assay | ≥95% |
| density | 2.24 g/mL at 25 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?