• USA Home
  • CRM846 - Pesticides - Loamy Sand 1

EMAIL THIS PAGE TO A FRIEND
CRM846 Sigma-Aldrich

Pesticides - Loamy Sand 1

certified reference material, pkg of 50 g, This product has been replaced by product CRM860-50G

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Environmental Matrix CRMs (RTC),
grade   certified reference material
packaging   pkg of 50 g
mfr. no.   RTC, CRM846-050
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
pH   7.12
format   matrix material

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis. Download your certificate of analysis at: http://www.sigma-aldrich.com.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

Aldrin SDS    
α-1,2,3,4,5,6-Hexachlorocyclohexane SDS    
β-BHC SDS    
δ-BHC SDS    
Lindane SDS    
α-Chlordane SDS    
1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane SDS    
1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene SDS    
4,4′-DDT SDS    
Dieldrin SDS    
Endosulfan sulfate SDS    
Endrin SDS    
Endrin aldehyde SDS    
Heptachlor SDS    
Methoxychlor SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?