• USA Home
  • D0922 - trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate

EMAIL THIS PAGE TO A FRIEND
D0922 Sigma-Aldrich

trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate

≥99% (titration)

Synonym: 1,2-Cyclohexanedinitrilotetraacetic acid, Chel-CD, DCTA

  • CAS Number 125572-95-4

  • Empirical Formula (Hill Notation) C14H22N2O8 · H2O

  • Molecular Weight 364.35

  •  Beilstein Registry Number 3222748

  •  EC Number 236-308-9

  •  MDL number MFCD00149243

  •  PubChem Substance ID 329798651

Purchase

Properties

Related Categories Cell Biology, D, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   VASZYFIKPKYGNC-DHTOPLTISA-N
assay   ≥99% (titration)
mp   213-216 °C(lit.)

Description

Legal Information

Chel is a trademark of Ciba-Geigy AG

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?