• USA Home
  • D3792 - 7-Deacetyl-1-deoxyforskolin from Coleus forskohlii

EMAIL THIS PAGE TO A FRIEND
D3792 Sigma-Aldrich

7-Deacetyl-1-deoxyforskolin from Coleus forskohlii

≥95%

Synonym: 6β,7β,9α-Trihydroxy-8,13-epoxy-labd-14-en-11-one

Purchase

Properties

InChI Key   BEXJRVNGEWHUJR-FTFGDVISSA-N
assay   ≥95%
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   −20°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?