• USA Home
  • D4042 - 4-(p-Dihexadecylaminostyryl)-N-methylquinolinium iodide

EMAIL THIS PAGE TO A FRIEND
D4042 Sigma-Aldrich

4-(p-Dihexadecylaminostyryl)-N-methylquinolinium iodide

≥95% (TLC)

Synonym: DiQ

Purchase

Properties

Related Categories Cell Biology, D, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   QLUOOALDIMFNGU-BYSMZXLCSA-N
assay   ≥95% (TLC)
storage temp.   −20°C

Description

Application

Membrane charge-shift probe

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?