Synonym: Diatomaceous earth, Diatomaceous silica, Kieselguhr
CAS Number 61790-53-2
MDL number MFCD00132803
PubChem Substance ID 24893841
Popular Documents: Specification Sheet (PDF)
| assay | ~90% SiO2 basis |
| form | powder |
| color | white to light gray |
Highly insoluble, inert material for use as a filtration aid.
1, 5 kg in poly drum
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?