• USA Home
  • D6628 - Dihydroergotamine (+)-tartrate salt

EMAIL THIS PAGE TO A FRIEND
D6628 Sigma-Aldrich

Dihydroergotamine (+)-tartrate salt

analytical standard

  • CAS Number 5989-77-5

  • Linear Formula (C33H37N5O5)2 · C4H6O6

  • Molecular Weight 1317.44

  •  EC Number 227-816-1

Purchase

Description

Biochem/physiol Actions

Agonist at vascular serotonin receptors; partial agonist at α-adrenergic and dopamine D2 receptors; vasoconstrictor.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
KE8000000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?