• USA Home
  • D7037 - Dextran sulfate sodium salt from Leuconostoc spp.

EMAIL THIS PAGE TO A FRIEND
D7037 Sigma-Aldrich

Dextran sulfate sodium salt from Leuconostoc spp.

average mol wt 5,000

Purchase

Properties

Related Categories Carbohydrates, Core Bioreagents, Research Essentials More...
mol wt   average mol wt 5,000
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
HH9290000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

D6924 Dextran sulfate sodium salt from Leuconostoc spp., average mol wt 9,000-20,000

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?