EMAIL THIS PAGE TO A FRIEND
E0399 Sigma-Aldrich

Edetate disodium

meets USP testing specifications

Synonym: Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Sequestrene Na2

  • CAS Number 6381-92-6

  • Empirical Formula (Hill Notation) C10H14N2Na2O8 · 2H2O

  • Molecular Weight 372.24

  •  Beilstein Registry Number 3900609

  •  EC Number 205-358-3

  •  PubChem Substance ID 24894368

Purchase

Description

Application

Chelator of divalent cations. Inhibits enzymes, such as metalloproteases, that require divalent cations for activity.

Other Notes

This product has been replaced by 27285-Sigma-Aldrich | Ethylenediaminetetraacetic acid disodium salt dihydrate meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%

Price and Availability


Filter out uncertainty.

All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory Tract
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AH4410000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?