EMAIL THIS PAGE TO A FRIEND
E7023 Sigma-Aldrich

Ethyl alcohol, Pure

200 proof, for molecular biology

Synonym: Ethanol, absolute alcohol, non-denatured ethanol

Purchase

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

Application

Suitable for use in the precipitation of nucleic acids

Biochem/physiol Actions

Positive allosteric modulator of GABAA receptors, and negative allosteric modulator of NMDA glutamate receptors.

General description

Prices include Federal Tax.

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Sales restrictions may apply

Packaging

1, 4, 4×4 L in glass bottle

500, 6×500 mL in glass bottle

Price and Availability


OEM & Contract Manufacturing Services Available
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1170 3 / PGII
WGK Germany 
1
RTECS 
KQ6300000
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C
Protocols & Articles

Articles

Recombinant Protein Expression in Saccharomyces cerevisiae

Expressing recombinant proteins in yeast has many advantages, simple protocols, use of basic reagents, appropriate processing of eukaryotic proteins, and scalability. Sigma® Life Science is an essent...
Keywords: Cell disruption, Gene expression, transformation

Protocols

Animal Tissue DNA-Extraction & WGA Amplification Protocol

Animal tissue is a common source of material when performing genetic analysis. The protocol below is a simple method of extracting DNA from the animal sample. Once the DNA has been isolated, it can t...

Blood Card - Extraction & Amplification WGA Protocol

Blood cards provide the convenience of archiving small volumes of blood. However, many times genomic DNA from these samples is limited, which may hinder the researcher’s ability to perform downstream...

Buccal DNA Extraction & WGA Amplification Protocol

This protocol provides a simple and convenient method to isolate, amplify, and purify genomic DNA from buccal swabs. Buccal swabs are a convenient method of acquiring a DNA sample. Once the DNA is is...

Chromatin Immunoprecipitation Kit CHP1 Protocol

Product Description Storage/Stability Workflow Preparation Instructions Procedure Analysis Troubleshooting Guide Frequently Asked Questions Materials References Poster & Application Note

Experienced User Protocol | GenElut Mammalian Genomic DNA Miniprep Kit

For experienced users, an abbreviated protocol is listed below. For data, images, troubleshooting and other information, please see the full length protocol page.
Keywords: PAGE

Extraction & Amplification of whole blood using WGA-Protocol

Whole blood is a common source of material used to perform genetic analysis. Many times genomic DNA isolated from whole blood samples is of low yield. This can hinder the researcher’s ability to perf...

GenElute 96 Well Blood Genomic DNA Purification Kit Protocol

The starting materials are lysed at room temperature in a lysis buffer which contains chaotropic salts in the presence of Proteinase K. The addition of ethanol ensures the appropriate conditions to b...
Keywords: Buffers, Cell disruption, Centrifugation, DNA purification, Digestions, Evaporation, Precipitation, Purification, Respiratory

GenElute 96 Well Tissue Genomic DNA Purification Kit Protocol

The starting material is lysed by incubation in a solution containing SDS and Proteinase K. Once lysis is complete, chaotropic salts are then added to further promote solubilization of macromolecules...
Keywords: Buffers, Cell disruption, Centrifugation, DNA purification, Evaporation, Polymerase chain reaction, Purification, Respiratory

GenElute Bacterial Genomic DNA Kit Protocol

Sigma's GenElute™ Bacterial Genomic DNA Kit provides a simple and convenient way to isolate pure DNA from a variety of cultured bacteria. The kit contains all of the reagents needed to isolate and pu...
Keywords: Amplification, Anticoagulants, Cell disruption, Centrifugation, Degradations, Digestions, Evaporation, Molecular biology, Nucleic acid denaturation, Phase transitions, Polymerase chain reaction, Precipitation

GenElute Mammalian Genomic DNA Miniprep Kit Protocol

Sigma’s GenElute Mammalian Genomic DNA Miniprep Kit provides a simple and convenient way to isolate pure genomic DNA from a variety of cultured cells, tissues (including rodent tails), and fresh whol...
Keywords: AGE, Amplification, Anticoagulants, Cell disruption, Centrifugation, DNA purification, Degradations, Digestions, Electrophoresis, Evaporation, Gel electrophoresis, Molecular biology, Nucleic acid denaturation, Phase transitions, Polymerase chain reaction, Precipitation, Purification, Sample preparations, Sequencing

GenElute Plant Genomic DNA Miniprep Kit Protocol (G2N10, G2N70, G2N350)

Several micrograms of DNA can be obtained from up to 100 mg of fresh tissue or 10 mg of freeze-dried material in less than an hour. The purified DNA is greater than 20 kb in length and can be used in...
Keywords: AGE, Amplification, Cell disruption, Centrifugation, Electrophoresis, Evaporation, Filtration, Gel electrophoresis, Molecular biology, Phase transitions, Polymerase chain reaction, Precipitation, Purification

GenElute™ Blood Genomic DNA Kit Protocol (NA2010, NA2020)

Sigma’s GenElute™ Blood Genomic DNA Kit provides a simple and convenient way to isolate pure genomic DNA from fresh or aged (older than 24 hours) whole blood. The kit combines the advantages of silic...
Keywords: AGE, Addition reactions, Amplification, Anticoagulants, Cell disruption, Centrifugation, Digestions, Electrophoresis, Evaporation, Gel electrophoresis, Molecular biology, Nucleic acid denaturation, Phase transitions, Polymerase chain reaction, Precipitation, Purification, Sequencing

MystiCq® MicroRNA® Quantitation System

Technology Overview Workflow Background Protocols miRNA isolation cDNA synthesis miRNA qPCR Advanced information Product selection guides Troubleshooting
Keywords: AGE, Amplification, Buffers, Cell disruption, Centrifugation, Degradations, Diagnostic, Digestions, Diseases, Electrophoresis, Gel electrophoresis, Gene expression, Homogenization, Microarray Analysis, Molecular biology, Nucleic acid annealing, Nucleic acid denaturation, Polymerase chain reaction, Polymerase chain reaction - quantitative, Precipitation, Purification, RNA purification, Sample preparations, Separation, Solvents, Titrations

Plant DNA extraction & WGA Amplification-protocol

Extracting DNA from plant tissue is a complicated process due to the tough cell wall that surrounds most plant cells. Genomic DNA from plant material can be damaged during the extraction process, res...

Plasma Serum DNA extraction & WGA Amplification Protocol

Whole genome amplification (WGA) of plasma and serum DNA presents a unique challenge due to the small amount of nucleic acid in such samples. One must use a robust DNA purification scheme capable of ...

Saliva DNA Extraction & WGA Amplification Protocol

Whole Genome Amplification can be performed on DNA extracted in many ways. Sigma-Aldrich offers many products for DNA extraction including the GenElute™ Blood Genomic DNA Kit (NA2010), GenElute Mamma...

Related Content

Immunohistochemistry Procedures

Immunohistochemical staining is a valuable tool for detecting specific antigens in tissues. In order to perform the standard staining procedure, first the tissue section has to be deparaffinized and ...
Keywords: Digestions, Immunohistochemistry, Solvents

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

also commonly purchased with this product

Product #

Description

Add to Cart

Z350370 Carestream® Kodak® BioMax® MR film, size 8 in. (20 cm) × 10 in. (25 cm)
59304 2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
C2432 Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
I9516 2-Propanol, BioReagent, for molecular biology, ≥99.5%

recommended

Product #

Description

Add to Cart

CLS3401 Corning® Transwell® polycarbonate membrane cell culture inserts, 12 mm Transwell with 0.4 μm pore polycarbonate membrane insert, TC-treated, w/ lid, sterile, 48/cs

suggested gloves for splash protection

Product #

Description

Add to Cart

Z677353 Dermatril P nitrile gloves, size XS
Z677388 Dermatril P nitrile gloves, size M
Z677396 Dermatril P nitrile gloves, size L
Z677418 Dermatril P nitrile gloves, size XL

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?