• USA Home
  • F6672 - Fraser secondary enrichment broth base

EMAIL THIS PAGE TO A FRIEND
F6672 Sigma-Aldrich

Fraser secondary enrichment broth base

for microbiology

Purchase

Description

Components

Ingredients (g/L)
Proteose peptone, 5.00
Casein enzymic hydrolysate, 5.00
Yeast extract, 5.00
Beef extract, 5.00
Sodium chloride, 20.00
Lithium chloride, 3.00
Disodium phosphate, 12.00
Monopotassium phosphate, 1.35
Esculin, 1.00
Ferric ammonium citrate, 0.50

Application

Recommended for isolation, cultivation, and enrichment of Listeria Monocytogenes.

Preparation Note

Supplement: 1 vial/l Fraser enrichment supplement (Cat. No. F2674)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Articles

Listeria a Survivor

Listeriosis is a serious infection caused by Listeria monocytogenes. In recent years it has been recognized that Listeria is an important public health problem. The disease affects primarily people o...
Jvo Siegrist
Microbiology Focus Edition 2.2
Keywords: AGE, Antibiotics, Diseases, Fermentation, Food Safety, Gastrointestinal, Indicators, Microbiology, Nutrition, Purification

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?