EMAIL THIS PAGE TO A FRIEND
F7923 Sigma-Aldrich

Ferrous fumarate

meets USP testing specifications

Synonym: Ferrous fumarate, Iron(II) fumarate

Purchase

Properties

Related Categories Analytical/Chromatography, Pharmacopoeia, Pharmacopoeia A-Z More...
InChI Key   PMVSDNDAUGGCCE-TYYBGVCCSA-L
suitability   meets USP testing specifications

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
LT1950000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?