• USA Home
  • G9511 - Glycophorin from blood type B negative

EMAIL THIS PAGE TO A FRIEND
G9511 Sigma-Aldrich

Glycophorin from blood type B negative

lyophilized powder, Predominantly Glycophorin A

Purchase

Properties

form   lyophilized powder
solubility   H2O: soluble, clear
storage temp.   2-8°C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?