• USA Home
  • G9539 - FreAlagin M gelatin

EMAIL THIS PAGE TO A FRIEND
G9539 Sigma-Aldrich

FreAlagin M gelatin

lyophilized powder

Synonym: Gelatin from porcine skin

Purchase

Properties

sterility   aseptically processed
form   lyophilized powder
mol wt   mol wt 200-10,000
impurities   endotoxin, free
storage temp.   2-8°C

Description

Components

Gelatin is a heterogeneous mixture of water-soluble proteins of high average molecular masses, present in collagen. Proteins are extracted by boiling the relevant skin, tendons, ligaments, bones, etc. in water. Type A gelatin is derived from acid-cured tissue. Type B is derived from lime-cured tissue.

Application

Exhibits a higher level of blocking activity than FreAlagin type R.

General description

FreAlagin gelatins are non-antigenic and low allergenic. They are recommended for use as stabilizers or blocking reagents.

Legal Information

FreAlagin is a trademark of Miyagi Chemical Industries Co., Ltd.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
LX8580000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?