• USA Home
  • H0777 - Heparin sodium salt from bovine intestinal mucosa

EMAIL THIS PAGE TO A FRIEND
H0777 Sigma-Aldrich

Heparin sodium salt from bovine intestinal mucosa

≥140 USP units/mg

Purchase

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
MI0850000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Glycosaminoglycan Sulfation and Signaling

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units. The disaccharide units that are the basis of all glycosaminoglycans (except keratan) are a combination...
Vicki Caligur
BioFiles 2008, 3.10, 4.
Keywords: Adhesion, Aminations, Angiogenesis, Anticoagulants, Apoptosis, Atomic absorption spectroscopy, Cancer, Cell biology, Cell proliferation, Cell signaling, Cellular processes, Central Nervous System, Chromatography, Coagulation, Degradations, Enzyme-linked immunosorbent assay, Epimerizations, Gene expression, Genetic, Genomics, Growth factors, Help, High performance liquid chromatography, Inflammation, Ion-exchange chromatography, Ligands, Mass spectrometry, Methods, Microarray Analysis, Morphogenesis, Nuclear magnetic resonance spectroscopy, Phosphorylations, Polymerase chain reaction, Polymerization reactions, Proteomics, Reductive aminations, Sequences, Spectroscopy, Sulfations, Transcription, Type, transformation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?