CAS Number 7722-84-1
Empirical Formula (Hill Notation) H2O2
Molecular Weight 34.01
Beilstein Registry Number 3587191
MDL number MFCD00011333
PubChem Substance ID 24895576
Popular Documents: Specification Sheet (PDF)
| Related Categories | Bioactive Small Molecules, Cell Biology, Chemical Synthesis, H, Oxidizing Agents, |
| vapor pressure | 23.3 mmHg ( 30 °C) |
| concentration | 30 wt. % in H2O |
| density | 1.11 g/mL at 25 °C |
| suitability | meets USP testing specifications |
| storage temp. | 2-8°C |
Live Chat and Frequently Asked Questions
are available for this Product.
1 L in poly bottle
500 mL in poly bottle
Stabilized
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?