• USA Home
  • H4396 - Histocryl embedding resin

EMAIL THIS PAGE TO A FRIEND
H4396 Sigma-Aldrich

Histocryl embedding resin

Purchase

Properties

Related Categories Cell Biology, G-H-I, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
storage temp.   2-8°C

Description

Application

Hydrophilic methacrylate resin
Benzoyl peroxide (B 5907) is specifically formulated as an initiator for this resin.

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3106A7 5.2
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?