• USA Home
  • H6535 - 6-Hexadecanoyl-2-(((2-(trimethylammonium)ethyl)methyl) amino)naphthalene chloride

EMAIL THIS PAGE TO A FRIEND
H6535 Sigma-Aldrich

6-Hexadecanoyl-2-(((2-(trimethylammonium)ethyl)methyl) amino)naphthalene chloride

solid

Synonym: Ethanaminium, N,N,N-trimethyl-2-(methyl-(6-(1-oxohexadecyl)-2naphthalenyl) amino), chloride, Patman

  • CAS Number 87393-54-2

  • Empirical Formula (Hill Notation) C32H53ClN2O

  • Molecular Weight 517.23

  •  MDL number MFCD00467561

Purchase

Properties

Related Categories Cell Biology, G-H-I, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   HEANZWXEJRRYTD-UHFFFAOYSA-M
form   solid

Description

Application

Polarity sensitive dye. Has been shown to conjugate with proteins in bilayer protein studies.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?